
2-Chloro-5-nitro-1,4-phenylenediamine ≥95.0% (by GC, titration analysis)
Catalog # TCIAC1501-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:155.0 to 158.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:187.58 g/mol
EINECS:247-515-9
:GHS07
CAS Number:26196-45-2
Formula:C₆H₆ClN₃O₂
MDL Number:MFCD00190136