Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Cyanoethyl)-2-ethyl-4-methylimidazole ≥70% (by GC) (contains 5-methyl isomer)
Toxic
1-(2-Cyanoethyl)-2-ethyl-4-methylimidazole ≥70% (by GC) (contains 5-methyl isomer)
Catalog # TCIAC1486-25G
Supplier:  TCI
CAS Number:  
Toxic
1-(2-Cyanoethyl)-2-ethyl-4-methylimidazole ≥70% (by GC) (contains 5-methyl isomer)
Catalog # TCIAC1486-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 70.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :163.22 g/mol
EINECS:245-975-5
UN:3276
ADR:6.1,III
 :GHS06
CAS Number:23996-25-0
Formula:C₉H₁₃N₃
MDL Number:MFCD00051492