6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5617-70-9
:
Formula:C₈H₁₀O₄
MDL Number:MFCD00042796
:170.16 g/mol
EINECS:227-044-5
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:69.0 to 72.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together