D-Cysteine hydrochloride monohydrate ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:207121-46-8
Formula:C₃H₇NO₂S·HCl·H₂O
MDL Number:MFCD00150051
:175.64 g/mol (157.62 anhy)
EINECS:251-043-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (Iodometric Titration):min. 99.0 % (calcd.on dried substance)
- Specific rotation [a]20/D:-5.0 to -7.0 deg (C=8, 1mol/L HCl)
- Solubility in Water:almost transparency
- Drying loss:8.0 to 11.0 %
Specification Test Results
Frequently Bought Together