Order Entry
Netherlands
ContactUsLinkComponent
1-Chloro-9,10-bis(phenylethynyl)anthracene ≥96.0% (by HPLC)
1-Chloro-9,10-bis(phenylethynyl)anthracene ≥96.0% (by HPLC)
Supplier:  TCI
Certificates
1-Chloro-9,10-bis(phenylethynyl)anthracene ≥96.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 96.0 area%

Specification Test Results

Related Information

Frequently Bought Together