

Chlorhexidine dihydrochloride ≥96.0% (by titrimetric analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:3697-42-5
:
Formule:C₂₂H₃₀Cl₂N₁₀
MDL-nummer:MFCD00068998
:578.37 g/mol
Merck index:14,02091
EINECS:223-026-6
UN:3077
ADR:9,III
:GHS07
:GHS09
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Solubility in Formic acid:almost transparency
Specificatie testresultaten
Frequently Bought Together