
(2-Carboxyethyl)dimethylsulphonium bromide ≥98.0% (by titrimetric analysis)
TCIAC1241-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Water:almost transparency
:
:215.11 g/mol
:GHS07
:20986-22-5
:C₅H₁₁BrO₂S
:MFCD00142887