
(2-Carboxyethyl)dimethylsulphonium chloride ≥98.0% (by titrimetric analysis)
TCIAC1240-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
:
:170.65 g/mol
:GHS07
:4337-33-1
:C₅H₁₁ClO₂S
:MFCD00142888