1,4-Cyclohexanedimethanol diisovalerate (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAC1135-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
Specification Test Results
About this item
:
:312.45 g/mol
CAS Number:377085-57-9
Formula:C₁₈H₃₂O₄
MDL Number:MFCD00060061