
1-(3-Chloropropyl)theobromine ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAC1103-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:144.0 to 148.0 deg-C
Specification Test Results
About this item
:
:256.69 g/mol
:GHS07
CAS Number:74409-52-2
Formula:C₁₀H₁₃ClN₄O₂
MDL Number:MFCD00059926