Order Entry
Netherlands
ContactUsLinkComponent
Isopropyl citrate (mixture) ≥97.0% (by GC)
Isopropyl citrate (mixture) ≥97.0% (by GC)
Catalog # TCIAC1030-25G
Supplier:  TCI
CAS Number:  
Isopropyl citrate (mixture) ≥97.0% (by GC)
Catalog # TCIAC1030-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow cloudy liquid
  • Purity (GC):
    min. 97.0 % (mixture)

Specification Test Results

About this item

 :
CAS Number:39413-05-3
MDL Number:MFCD00060079