(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0996-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:127.0 to 131.0 deg-C
- Specific rotation [a]20/D:-11.0 to -12.0 deg (C=8, 1mol/L HCl)
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:185.666 g/mol
Storage Temperature:Refrigerator
EINECS:212-779-6
CAS Number:868-59-7
Formula:C₅H₁₁NO₂S·HCl
MDL Number:MFCD00012631