Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Chloro-4-pyridyl)-3-phenylurea ≥98.0% (by titrimetric analysis)
Irritant
Health
Environment
1-(2-Chloro-4-pyridyl)-3-phenylurea ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0926-5G
Supplier:  TCI
CAS Number:  
Irritant
Health
Environment
1-(2-Chloro-4-pyridyl)-3-phenylurea ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0926-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    168.0 to 172.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :247.68 g/mol
Storage Temperature:Refrigerator
UN:3077
ADR:9,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:68157-60-8
Formula:C₁₂H₁₀ClN₃O
MDL Number:MFCD00059898