cis-3-Hexen-1-ylhexanoate ≥98.0% (by GC)
Catalog # TCIAC0917-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
Flash Pt:116°C
:198.31 g/mol
EINECS:250-661-6
CAS Number:31501-11-8
Formula:C₁₂H₂₂O₂
MDL Number:MFCD00036552