
Cyanoacetohydrazide ≥97.0% (by titrimetric analysis)
Catalog # TCIAC0800-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (Bromination Method):min. 97.0 %
- Melting point:107.0 to 110.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:99.09 g/mol
Merck Index:14,02680
EINECS:205-437-2
UN:2811
ADR:6.1,III
:GHS06
CAS Number:140-87-4
Formula:C₃H₅N₃O
MDL Number:MFCD00007611