Nɑ-Carbobenzoxy-L-arginine ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1234-35-1
:
Formula:C₁₄H₂₀N₄O₄
MDL Number:MFCD00001762
:308.34 g/mol
EINECS:214-973-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
- Specific rotation [a]20/D:-9.0 to -11.0 deg (C=5.5, 0.2mol/L HCl)
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together