Cholesterol formate ≥96.0% (by GC, titration analysis)
TCIAC0671-10G
: TCI
:
- Pk:10 g
- Seq:3236
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Ester Value):min. 96.0 %
- Melting point:94.0 to 99.0 deg-C
- Specific rotation [a]20/D:-45.0 to -57.0 deg (C=2, CHCl3)
:
:414.67 g/mol
:224-416-9
:4351-55-7
:C₂₈H₄₆O₂
:MFCD00046243