Order Entry
Netherlands
ContactUsLinkComponent
Tripropyl citrate ≥97.0% (by titrimetric analysis)
Tripropyl citrate ≥97.0% (by titrimetric analysis)
Catalog # TCIAC0601-25G
Supplier:  TCI
CAS Number:  
Tripropyl citrate ≥97.0% (by titrimetric analysis)
Catalog # TCIAC0601-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (Ester Value):
    min. 97.0 %

Specification Test Results

About this item

 :
 :318.37 g/mol
Boiling Pt:198°C
EINECS:216-450-8
CAS Number:1587-21-9
Formula:C₁₅H₂₆O₇
MDL Number:MFCD00059407