Order Entry
Netherlands
ContactUsLinkComponent
L-Cystine dihydrochloride ≥98.0% (by HPLC)
L-Cystine dihydrochloride ≥98.0% (by HPLC)
Catalog # TCIAC0520-25G
Supplier:  TCI
CAS Number:  
L-Cystine dihydrochloride ≥98.0% (by HPLC)
Catalog # TCIAC0520-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    -163 to -173 deg (C=1, 1mol/L HCl)
  • Solubility in 1mol/L HCl:
    almost transparency

Specification Test Results

About this item

 :
 :313.23 g/mol
EINECS:250-391-9
CAS Number:30925-07-6
Formula:C₆H₁₂N₂O₄S₂·2HCl
MDL Number:MFCD00070399