Order Entry
Netherlands
ContactUsLinkComponent
DL-Cysteine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
 
DL-Cysteine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
  TCIAC0516-25G
 :  TCI
 :  
 
DL-Cysteine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
  TCIAC0516-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (Iodometric Titration):
    min. 98.0% (calcd.on dried substance)
  • Solubility in Water:
    almost transparency
  • Drying loss:
    8.5 to 12.0 %

 

 

 :
 :175.63 g/mol
 :14,02781
 :GHS07
 :116797-51-4
 :96998-61-7
 :C₃H₇NO₂S·HCl·H₂O
 :MFCD00151030