Corticosterone ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:50-22-6
:
Formula:C₂₁H₃₀O₄
MDL Number:MFCD00037715
Merck Index:14,02538
:346.5 g/mol
EINECS:200-019-6
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:181.0 to 184.0 deg-C
- Specific rotation [a]20/D:+221 to +226 deg (C=1, EtOH)
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together