Order Entry
Netherlands
ContactUsLinkComponent
cis,trans-5,9-Cyclododecadiene-cis-1,2-diol ≥98.0% (by GC)
cis,trans-5,9-Cyclododecadiene-cis-1,2-diol ≥98.0% (by GC)
Supplier:  TCI
Certificates
cis,trans-5,9-Cyclododecadiene-cis-1,2-diol ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    156.0 to 161.0 deg-C

Specification Test Results

Frequently Bought Together