
Cinchonidine dihydrochloride ≥98.0% (by titrimetric analysis)
TCIAC0348-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:-140 to -150 deg (C=0.4, H2O)
- Solubility in Water:very faint turbidity
:
:367.31 g/mol
:246-139-2
:GHS07
:24302-67-8
:C₁₉H₂₂N₂O
:MFCD00082451