
Sodium hydrogen-4-chlorophthalate ≥70% (by HPLC) (contains isomer and Phthalic Acid)
Catalog # TCIAC0265-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 70.0 area%
- Neutralization Value:232 to 238
Specification Test Results
About this item
:
:222.56 g/mol
EINECS:259-963-2
UN:2811
ADR:6.1,III
:GHS06
CAS Number:56047-23-5
Formula:C₈H₅ClO₄
MDL Number:MFCD00002495