N,N-Bis(4-bromophenyl)-[1,1':4',1''-terphenyl]-4-amine ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:2205054-16-4
:
Formula:C₃₀H₂₁Br₂N
:555.31 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:193.0 to 197.0 deg-C
Specification Test Results
Frequently Bought Together