N-[4-(tert-Butyl)phenyl]-9,9-dimethyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)
Catalogus # TCIAB6084-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:114.0 to 118.0 deg-C
Specification Test Results
About this item
:
:341.50 g/mol
CAS Number:944418-46-6
Formula:C₂₅H₂₇N
MDL Number:MFCD32068070