N-([1,1'-Biphenyl]-4-yl)phenanthren-9-amine ≥98.0% (by GC)
Catalog # TCIAB6083-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:173.0 to 177.0 deg-C
Specification Test Results
About this item
:
:345.45 g/mol
CAS Number:443965-64-8
Formula:C₂₆H₁₉N