Order Entry
Netherlands
ContactUsLinkComponent
 
5,5'-Bis(triisopropoxysilyl)-2,2'-bipyridine ≥95.0% (by GC)
  TCIAB6049-1G
 :  TCI
 :  
5,5'-Bis(triisopropoxysilyl)-2,2'-bipyridine ≥95.0% (by GC)
  TCIAB6049-1G
 :  TCI
 :  

 

  • Pk:
    1 g/mol
  • Seq:
    9220

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :564.87 g/mol
 :1569022-20-3
 :C₂₈H₄₈N₂O₆Si₂