N,N'-Bis(2-octyldodecyl)-6,6'-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoindigo
Catalog # TCIAB5994-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Red to Dark red to Brown powder to crystal
- Melting point:104.0 to 108.0 deg-C
- Elemental analysis (Nitrogen):2.40 to 2.80 %
Specification Test Results
About this item
:
:1075.27 g/mol
CAS Number:1563062-80-5
Formula:C₆₈H₁₁₂B₂N₂O₆