N,N'-Bis(2-ethylhexyl)-6,6'-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoindigo
Catalog # TCIAB5993-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Red to Dark red to Brown powder to crystal
- Melting point:160.0 to 164.0 deg-C
- Elemental analysis (Nitrogen):3.60 to 4.00 %
Specification Test Results
About this item
:
:738.62 g/mol
CAS Number:1821433-54-8
Formula:C₄₄H₆₄B₂N₂O₆