Order Entry
Netherlands
ContactUsLinkComponent
4-Bromo-4'-(4-bromophenyl)-3',5',6'-triphenyl-1,1':2',1''-terphenyl ≥98.0% (by HPLC)
4-Bromo-4'-(4-bromophenyl)-3',5',6'-triphenyl-1,1':2',1''-terphenyl ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
4-Bromo-4'-(4-bromophenyl)-3',5',6'-triphenyl-1,1':2',1''-terphenyl ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

Frequently Bought Together