Order Entry
Netherlands
ContactUsLinkComponent
 
1,4-Bis(3-isocyanopropyl)piperazine ≥97.0% (by HPLC)
Catalog # TCIAB5973-1G
Supplier:  TCI
CAS Number:  
1,4-Bis(3-isocyanopropyl)piperazine ≥97.0% (by HPLC)
Catalog # TCIAB5973-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Melting point:
    81.0 to 85.0 deg-C

Specification Test Results

About this item

 :
 :220.32 g/mol
CAS Number:51641-96-4
Formula:C₁₂H₂₀N₄
MDL Number:MFCD06200487