Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate ≥95.0% (by GC)
Irritant
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate ≥95.0% (by GC)
Supplier:  TCI
Certificates
Irritant
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Frequently Bought Together