2'-Bromospiro[fluorene-9,9'-xanthene] ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1477458-14-2
:
Formula:C₂₅H₁₅BrO
MDL Number:MFCD31814409
:411.30 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:195.0 to 199.0 deg-C
Specification Test Results
Frequently Bought Together