
2,4-Bis(phenylsulphonyl)phenol ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:177325-75-6
:
Formula:C₁₈H₁₄O₅S₂
MDL Number:MFCD29072767
:374.43 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Melting point:156.0 to 160.0 deg-C
Specification Test Results
Frequently Bought Together