Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate ≥98.0% (by titrimetric analysis)
tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    140.0 to 144.0 deg-C

Specification Test Results

Frequently Bought Together