
1,3-Bis[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]-1,1,3,3-tetramethyldisiloxane ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18724-32-8
:
Formula:C₂₀H₃₈O₃Si₂
MDL Number:MFCD03262840
:382.69 g/mol
Boiling Pt:195°C
Flash Pt:200°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together