(1R,2R)-1,2-Bis(2,4,6-trimethoxyphenyl)ethylenediamine dihydrochloride ≥93.0% (by HPLC)
TCIAB5916-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 93.0 area%
- Specific rotation [a]20/D:+71.0 to +75.0 deg (C=1, MeOH)
:
:465.37 g/mol
:1055301-18-2
:C₂₀H₂₈N₂O₆·₂HCl
:MFCD09953700