Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl(2-iodoethoxy)dimethylsilane ≥97.0% (by GC)
Irritant
tert-Butyl(2-iodoethoxy)dimethylsilane ≥97.0% (by GC)
Catalog # TCIAB5885-1G
Supplier:  TCI
CAS Number:  
Irritant
tert-Butyl(2-iodoethoxy)dimethylsilane ≥97.0% (by GC)
Catalog # TCIAB5885-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Yellow to Orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :286.23 g/mol
Boiling Pt:65°C
 :GHS07
CAS Number:101166-65-8
Formula:C₈H₁₉IOSi
MDL Number:MFCD09909940