Order Entry
Netherlands
ContactUsLinkComponent
1,2-Bis(4-bromophenyl)ethane ≥98.0% (by GC)
1,2-Bis(4-bromophenyl)ethane ≥98.0% (by GC)
Supplier:  TCI
Certificates
1,2-Bis(4-bromophenyl)ethane ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    116.0 to 120.0 deg-C

Specification Test Results

Frequently Bought Together