
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:170011-57-1
:
Formula:C₁₆H₂₄N₂O₂
MDL Number:MFCD05861405
:276.3740 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:117.0 to 121.0 deg-C
Specification Test Results
Frequently Bought Together