Order Entry
Netherlands
ContactUsLinkComponent
 
 
(R)-3-(Bromomethyl)heptane ≥95.0% (by GC)
 :  TCI
 
 
(R)-3-(Bromomethyl)heptane ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %
  • Optical purity (GC):
    min. 95.0 ee%

 

Frequently Bought Together