

[4,8-Bis[5-(2-ethylhexyl)thieno[3,2-b]thiophen-2-yl]benzo[1,2-b:4,5-b']dithiophene-2,6-diyl]bis(trimethylstannane) ≥97.0% (by HPLC)
Catalog # TCIAB5768-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:1016.73 g/mol
UN:3146
ADR:6.1,I
:GHS06
:GHS09
CAS Number:1444200-40-1
Formula:C₄₄H₅₈S₆Sn₂