N-([1,1'-Biphenyl]-4-yl)dibenzo[b,d]thiophen-4-amine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1448185-87-2
:
Formula:C₂₄H₁₇NS
MDL Number:MFCD28138080
:351.47 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:186.0 to 190.0 deg-C
Specification Test Results
Frequently Bought Together