Order Entry
Netherlands
ContactUsLinkComponent
9-([1,1'-Biphenyl]-3-yl)-10-bromoanthracene ≥98.0% (by GC)
9-([1,1'-Biphenyl]-3-yl)-10-bromoanthracene ≥98.0% (by GC)
Supplier:  TCI
Certificates
9-([1,1'-Biphenyl]-3-yl)-10-bromoanthracene ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together