tert-Butyl (3R,4S)-2-Oxo-4-phenyl-3-[(triethylsilyl)oxy]azetidine-1-carboxylate ≥97.0% (by HPLC)
TCIAB5715-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Colorless to Light yellow clear liquid
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
:
:377.56 g/mol
:Freezer
:149198-47-0
:C₂₀H₃₁NO₄Si
:MFCD08273948