N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:127828-22-2
:
Formula:C₉H₂₀N₂O₃
MDL Number:MFCD12031501
:204.27 g/mol
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together