Order Entry
Netherlands
ContactUsLinkComponent
(11-Bromoundecyl)trichlorosilane ≥97.0% (by GC)
Corrosive
(11-Bromoundecyl)trichlorosilane ≥97.0% (by GC)
Catalog # TCIAB5680-1G
Supplier:  TCI
CAS Number:  
Corrosive
(11-Bromoundecyl)trichlorosilane ≥97.0% (by GC)
Catalog # TCIAB5680-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :368.64 g/mol
Boiling Pt:65°C
UN:2987
ADR:8,II
 :GHS05
CAS Number:79769-48-5
Formula:C₁₁H₂₂BrCl₃Si
MDL Number:MFCD00270977