Order Entry
Netherlands
ContactUsLinkComponent
3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane ≥95.0% (by GC)
Irritant
3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    175.0 to 179.0 deg-C

Specification Test Results

Frequently Bought Together