Order Entry
Netherlands
ContactUsLinkComponent
(1S,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥98.0% (by GC, titration analysis)
Irritant
(1S,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
(1S,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Optical purity (GC):
    min. 98.0 ee%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    115.0 to 119.0 deg-C

Specification Test Results

Frequently Bought Together